Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kallikrein 1 Antibody (105), PerCP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289474PCP
Description
Kallikrein 1 Monoclonal antibody specifically detects Kallikrein 1 in Human samples. It is validated for ELISASpecifications
Kallikrein 1 | |
Monoclonal | |
PerCP | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
105 | |
ELISA | |
EC 3.4.21, EC 3.4.21.35, glandular kallikrein 1, hK1, kallikrein 1, kallikrein 1, renal/pancreas/salivary, kallikrein serine protease 1, kallikrein-1, Kidney/pancreas/salivary gland kallikrein, Klk6, KLKR, Tissue kallikrein | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Kallikrein 1 (Accession#: NP_002248.1; Met 1-Ser 262). | |
0.1 mL | |
Cancer | |
3816 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction