Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kallikrein 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579562
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579562 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579562 Supplier Invitrogen™ Supplier No. PA579562
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat pancreas tissue, mouse pancreas tissue. IHC: Human Pancreatic Cancer Tissue.

Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in a cluster on chromosome 19. This protein is functionally conserved in its capacity to release the vasoactive peptide, Lys-bradykinin, from low molecular weight kininogen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kallikrein 1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene KLK1
Gene Accession No. P00756, P00758, P06870, P15947, P36373
Gene Alias 0610007D04Rik; 7S nerve growth factor gamma chain; 7S nerve growth factor gamma subunit; esterase B; Gamma-NGF; glandular kallikrein 1; glandular kallikrein K1; Glandular kallikrein K3; glandular kallikrein-7, submandibular/renal; hK1; Kal; KALA; KAL-B; kallikrein 1; Kallikrein 1 renal/pancreas/salivary; kallikrein 1, renal/pancreas/salivary; kallikrein 1-like peptidase; kallikrein 1-related peptidase b3; kallikrein 1-related peptidase C7; kallikrein 5; kallikrein 6; kallikrein 7; kallikrein renal/pancreas/salivary; kallikrein serine protease 1; kallikrein-1; Kallikrein-related protein K1; kidney/pancreas/salivary gland kallikrein; Klk1; Klk-1; Klk1b3; Klk1b6; Klk1c1; Klk1c7; Klk1l; Klk3; Klk-3; Klk5; Klk6; Klk-6; Klk7; Klk-7; KLKR; mGk-3; mGK-6; Nerve growth factor gamma chain; Nerve growth factor gamma chain 1; Nerve growth factor gamma chain 2; nerve growth factor gamma polypeptide; nerve growth factor, gamma; Ngfg; Pancreatic kallikrein; proteinase A; PS kallikrein; RATKALA; Renal kallikrein; RGK-1; RGK-7; rK-1; RSKG-7; Tissue kallikrein; tissue kallikrein-3; Tissue kallikrein-6; True kallikrein; True tissue kallikrein
Gene Symbols KLK1, Klk1b3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence in the middle region of human Kallikrein 1 (182-197aa NDECKKAHVQKVTDFM).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16612, 18050, 24523, 24594, 3816
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.