Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kallikrein 1 Recombinant Rabbit Monoclonal Antibody (105)
GREENER_CHOICE

Catalog No. PIMA530955
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530955 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530955 Supplier Invitrogen™ Supplier No. MA530955
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human KLK1/Kallikrein 1.

Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in a cluster on chromosome 19. This protein is functionally conserved in its capacity to release the vasoactive peptide, Lys-bradykinin, from low molecular weight kininogen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kallikrein 1
Applications ELISA
Classification Recombinant Monoclonal
Clone 105
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene KLK1
Gene Accession No. P06870
Gene Alias 0610007D04Rik; 7S nerve growth factor gamma chain; 7S nerve growth factor gamma subunit; esterase B; Gamma-NGF; glandular kallikrein 1; glandular kallikrein K1; Glandular kallikrein K3; glandular kallikrein-7, submandibular/renal; hK1; Kal; KALA; KAL-B; kallikrein 1; Kallikrein 1 renal/pancreas/salivary; kallikrein 1, renal/pancreas/salivary; kallikrein 1-like peptidase; kallikrein 1-related peptidase b3; kallikrein 1-related peptidase C7; kallikrein 5; kallikrein 6; kallikrein 7; kallikrein renal/pancreas/salivary; kallikrein serine protease 1; kallikrein-1; Kallikrein-related protein K1; kidney/pancreas/salivary gland kallikrein; Klk1; Klk-1; Klk1b3; Klk1b6; Klk1c1; Klk1c7; Klk1l; Klk3; Klk-3; Klk5; Klk6; Klk-6; Klk7; Klk-7; KLKR; mGk-3; mGK-6; Nerve growth factor gamma chain; Nerve growth factor gamma chain 1; Nerve growth factor gamma chain 2; nerve growth factor gamma polypeptide; nerve growth factor, gamma; Ngfg; Pancreatic kallikrein; proteinase A; PS kallikrein; RATKALA; Renal kallikrein; RGK-1; RGK-7; rK-1; RSKG-7; Tissue kallikrein; tissue kallikrein-3; Tissue kallikrein-6; True kallikrein; True tissue kallikrein
Gene Symbols KLK1
Host Species Rabbit
Immunogen Recombinant Human KLK1/Kallikrein 1 Protein (Met1-Ser262).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3816
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.