Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kallikrein 10 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP219971
Description
Kallikrein 10 Polyclonal specifically detects Kallikrein 10 in Human samples. It is validated for Western Blot.Specifications
Kallikrein 10 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
breast normal epithelial cell associated serine protease, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.74, Kallikrein 10, kallikrein-related peptidase 10, NES1kallikrein-10, Normal epithelial cell-specific 1, Protease serine-like 1, PRSSL1protease, serine-like, 1 | |
Rabbit | |
30 kDa | |
0.1 mL | |
Cell Cycle and Replication | |
5655 | |
Human | |
IgG |
Western Blot | |
0.38mg/mL | |
Western Blot 1:500-1:3000 | |
O43240 | |
KLK10 | |
Recombinant protein encompassing a sequence within the center region of human Kallikrein 10. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human. Species reactivity based on sequence identity: Dog (88%), Bovine (86%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction