Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kallikrein 11 Antibody (101), Janelia Fluor™ 549 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289629JF549
Description
Kallikrein 11 Monoclonal antibody specifically detects Kallikrein 11 in Human samples. It is validated for ELISA, ELISASpecifications
Kallikrein 11 | |
Monoclonal | |
Janelia Fluor 549 | |
EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, Hippostasin, hK11, kallikrein 11, kallikrein-related peptidase 11, MGC33060, PRSS20kallikrein-11, Serine protease 20, TLSPhippostasin, Trypsin-like protease | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Kallikrein 11 (NP_006844.1; Met1-Asn250). | |
0.1 mL | |
Cancer | |
11012 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
101 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction