Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kallikrein 11 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100429
Description
Kallikrein 11 Polyclonal specifically detects Kallikrein 11 in Human samples. It is validated for Western Blot.Specifications
Kallikrein 11 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
KLK11 | |
Residues 68-82 [YIVHLGQHNLQKEEG] of the 35 kDa human hippostasin/KLK11protein. Sequence identity to mouse and rat: 86%. | |
Affinity Purified | |
RUO | |
Primary | |
This is specific for Hippostasin/KLK11. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot | |
1 mg/mL | |
Western Blot 1:500-1:1000 | |
EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, Hippostasin, hK11, kallikrein 11, kallikrein-related peptidase 11, MGC33060, PRSS20kallikrein-11, Serine protease 20, TLSPhippostasin, Trypsin-like protease | |
Rabbit | |
31 kDa | |
0.1 mL | |
Cancer | |
11012 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction