Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kallikrein 11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA14312
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA14312 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA14312 Supplier Invitrogen™ Supplier No. PA14312
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-4312 detects the Hippostatin/KLK11 protein in human, mouse, and rat samples. PA1-4312 has been successfully used in Western blot procedures. By Western blot this antibody detects a ~38 kDa protein representing KLK11 in LNCaP cell lysate. PA1-4312 immunizing peptide corresponds to amino acid residues 68-82 of the 35 kDa human hippostasin/KLK11protein. There is 86% sequence identity to mouse and rat.

Hippostasin, also known as KLK11, is a kallikrein-like serine protease, which has two alternatively spliced isoforms, brain-type and prostate-type. Hippostasin plays a role in the prostate, including reproductive and/or tumorigenic functions. Elevated serum levels of KLK11 were found in 70% of women with ovarian cancer and in 60% of men with prostate cancer. Analysis of the KLK11 biomarker in serum may aid in the diagnosis and monitoring of ovarian and prostatic carcinoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kallikrein 11
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Klk11
Gene Accession No. Q9QYN3, Q9UBX7
Gene Alias 2310015I08Rik; Hippostasin; hK11; kallikrein 11; kallikrein related peptidase 11; kallikrein related-peptidase 11; Kallikrein11; kallikrein-11; Kallikrein-11 inactive chain 1; Kallikrein-11 inactive chain 2; kallikrein-related peptidase 11; KLK11; protease, serine, 20; Prss20; Serine protease 20; TLSP; Trypsin-like protease; UNQ649/PRO1279
Gene Symbols Klk11
Host Species Rabbit
Immunogen Synthetic peptide: Y(68) I V H L G Q H N L Q K E E G(82).
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11012, 292849, 56538
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.