Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kallikrein 11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528008
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528008 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528008 Supplier Invitrogen™ Supplier No. PA528008
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji , mouse brain. Predicted reactivity: Mouse (81%), Rat (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Hippostasin, also known as KLK11, is a kallikrein-like serine protease, which has two alternatively spliced isoforms, brain-type and prostate-type. Hippostasin plays a role in the prostate, including reproductive and/or tumorigenic functions. Elevated serum levels of KLK11 were found in 70% of women with ovarian cancer and in 60% of men with prostate cancer. Analysis of the KLK11 biomarker in serum may aid in the diagnosis and monitoring of ovarian and prostatic carcinoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kallikrein 11
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene Klk11
Gene Accession No. Q9QYN3, Q9UBX7
Gene Alias 2310015I08Rik; Hippostasin; hK11; kallikrein 11; kallikrein related peptidase 11; kallikrein related-peptidase 11; Kallikrein11; kallikrein-11; Kallikrein-11 inactive chain 1; Kallikrein-11 inactive chain 2; kallikrein-related peptidase 11; KLK11; protease, serine, 20; Prss20; Serine protease 20; TLSP; Trypsin-like protease; UNQ649/PRO1279
Gene Symbols Klk11
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Kallikrein 11. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11012, 56538
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.