Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kallikrein 11 Recombinant Rabbit Monoclonal Antibody (121)
GREENER_CHOICE

Catalog No. PIMA529377
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529377 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529377 Supplier Invitrogen™ Supplier No. MA529377
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human KLK-11/Kallikrein 11.

Hippostasin, also known as KLK11, is a kallikrein-like serine protease, which has two alternatively spliced isoforms, brain-type and prostate-type. Hippostasin plays a role in the prostate, including reproductive and/or tumorigenic functions. Elevated serum levels of KLK11 were found in 70% of women with ovarian cancer and in 60% of men with prostate cancer. Analysis of the KLK11 biomarker in serum may aid in the diagnosis and monitoring of ovarian and prostatic carcinoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kallikrein 11
Applications ELISA, Immunoprecipitation, Western Blot
Classification Recombinant Monoclonal
Clone 121
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Klk11
Gene Accession No. Q9UBX7
Gene Alias 2310015I08Rik; Hippostasin; hK11; kallikrein 11; kallikrein related peptidase 11; kallikrein related-peptidase 11; Kallikrein11; kallikrein-11; Kallikrein-11 inactive chain 1; Kallikrein-11 inactive chain 2; kallikrein-related peptidase 11; KLK11; protease, serine, 20; Prss20; Serine protease 20; TLSP; Trypsin-like protease; UNQ649/PRO1279
Gene Symbols Klk11
Host Species Rabbit
Immunogen Recombinant Human KLK-11 protein (Met1-Asn250).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11012
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.