Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kallikrein 12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA14237
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA14237 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA14237 Supplier Invitrogen™ Supplier No. PA14237
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-4237 detects Kallikrein-12 from human samples. PA1-4237 has been successfully used in Western blot procedures. PA1-4237 immunizing peptide corresponds to amino acid residues 236-248 from human Kallikrein-12.

Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in a cluster on chromosome 19. Alternate splicing of this gene results in three transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kallikrein 12
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene KLK12
Gene Accession No. Q9UKR0
Gene Alias 2310008B01Rik; AI324852; kallikrein 12; kallikrein related peptidase 12; kallikrein related-peptidase 12; kallikrein-12; Kallikrein-like protein 5; kallikrein-related peptidase 12; KLK12; KLK-12; KLKL5; KLK-L5; UNQ669/PRO1303
Gene Symbols KLK12
Host Species Rabbit
Immunogen Synthetic Peptide: K(236) Y V D W I R M I M R N N(248).
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 43849
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.