Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kallikrein 13 Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.50 - $499.50

Specifications

Antigen Kallikrein 13
Concentration 1 mg/mL
Dilution Western Blot 1:500- 1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018121
SDP
View Documents
Novus Biologicals
NB200139SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NB200139
SDP
View Documents
Novus Biologicals
NB200139
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

Kallikrein 13 Polyclonal specifically detects Kallikrein 13 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

Kallikrein 13
Western Blot 1:500- 1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Polyclonal
Rabbit
Human
Q9UKR3
26085
Residues 262-277 [IRKYETQQQKWLKGPQ] of the human KLK-L4 protein.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
31 kDa
1 mg/mL
ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Unconjugated
RUO
PBS with 0.02% Sodium Azide
DKFZp586J1923, EC 3.4.21, EC 3.4.21.-, kallikrein 13, Kallikrein-like protein 4, kallikrein-related peptidase 13, KLK-L4kallikrein-13, KLKL4kallikrein-like gene 4
KLK13
IgG
Affinity Purified
Reacts with residues 262-277 [IRKYETQQQKWLKGPQ] of the human KLK-L4 protein.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.