Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kallikrein 13 Monoclonal Antibody (10H10A2C4)
GREENER_CHOICE

Catalog No. PIMA530960
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530960 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530960 Supplier Invitrogen™ Supplier No. MA530960
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human KLK13/Kallikrein 13.

Kallikreins are a subgroup of serine proteases and are implicated in carcinogenesis. The novel KLK gene, KLK-L4, is expressed in a variety of tissues including prostate, salivary gland, breast, and testis. Its expression is regulated by steroid hormones in breast cancer cell lines may be involved in the pathogenesis and/or progression of breast cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kallikrein 13
Applications ELISA
Classification Monoclonal
Clone 10H10A2C4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene KLK13
Gene Accession No. Q9UKR3
Gene Alias DKFZp586J1923; Egfbp-2; kallikrein 13; kallikrein related peptidase 13; kallikrein related-peptidase 13; kallikrein-13; kallikrein-like gene 4; kallikrein-like protein 4; kallikrein-related peptidase 13; KLK13; KLK-13; KLKL4; KLK-L4; mGk-13
Gene Symbols KLK13
Host Species Mouse
Immunogen Recombinant Human KLK13 protein (Met1-Ile262).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26085
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.