Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kallikrein 14 Monoclonal Antibody (337903)
GREENER_CHOICE

Catalog No. PIMA524067
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524067 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524067 Supplier Invitrogen™ Supplier No. MA524067
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute at 0.5 mg/mL in sterile PBS.

Kallikreins are a subgroup of serine proteases and are implicated in carcinogenesis. A novel KLK gene, KLK12, is expressed in a variety of tissues including salivary gland, stomach, uterus, lung, thymus, prostate, colon, brain, thyroid, and trachea. It has applications as a novel cancer biomarker.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kallikrein 14
Applications Immunoprecipitation, Neutralization
Classification Monoclonal
Clone 337903
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Klk14
Gene Accession No. Q9P0G3
Gene Alias GK14; Glandular kallikrein KLK14; hK14; kallikrein 14; Kallikrein L6; kallikrein related peptidase 14; kallikrein related-peptidase 14; Kallikrein-14; kallikrein-like protein 6; kallikrein-related peptidase 14; KLK14; KLK-14; KLKL6; KLK-L6; mGK14
Gene Symbols Klk14
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Kallikrein 14 Gln19-Met248.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 43847
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.