Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kallikrein 14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588032
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588032 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588032 Supplier Invitrogen™ Supplier No. PA588032
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IIGGHTCTRS SQPWQAALLA GPRRRFLCGG ALLSGQWVIT AAHCGRPILQ VALGKHNLRR WEATQQVLRV VRQVTHPNYN SRTHDNDLML LQLQQPARIG RAVRPIEVTQ ACASPGTSCR VSGWGTISSP IARYPASLQC VNINISPDEV CQKAYPRTIT PGMVCAGVPQ GGKDSCQGDS GGPLVCRGQL QGLVSWGMER CALPGYPGVY TNLCKYRSWI EETMRDK; Positive Samples: Mouse stomach; Cellular Location: extracellular space.

Kallikreins are a subgroup of serine proteases and are implicated in carcinogenesis. A novel KLK gene, KLK12, is expressed in a variety of tissues including salivary gland, stomach, uterus, lung, thymus, prostate, colon, brain, thyroid, and trachea. It has applications as a novel cancer biomarker.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kallikrein 14
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Klk14
Gene Accession No. Q8CGR5
Gene Alias GK14; Glandular kallikrein KLK14; hK14; kallikrein 14; Kallikrein L6; kallikrein related peptidase 14; kallikrein related-peptidase 14; Kallikrein-14; kallikrein-like protein 6; kallikrein-related peptidase 14; KLK14; KLK-14; KLKL6; KLK-L6; mGK14
Gene Symbols Klk14
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 41-267 of human KLK14 (NP_0713292).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 317653
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.