Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Kallikrein 14 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA547354
Description
In sandwich ELISAs, less than 0.05% cross-reactivity with recombinant human Kallikrein 1, 3, 4, 5, 7, 8, 11, 12, and 13 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Kallikreins are a subgroup of serine proteases and are implicated in carcinogenesis. A novel KLK gene, KLK12, is expressed in a variety of tissues including salivary gland, stomach, uterus, lung, thymus, prostate, colon, brain, thyroid, and trachea. It has applications as a novel cancer biomarker.
Specifications
| Kallikrein 14 | |
| Polyclonal | |
| Unconjugated | |
| Klk14 | |
| GK14; Glandular kallikrein KLK14; hK14; kallikrein 14; Kallikrein L6; kallikrein related peptidase 14; kallikrein related-peptidase 14; Kallikrein-14; kallikrein-like protein 6; kallikrein-related peptidase 14; KLK14; KLK-14; KLKL6; KLK-L6; mGK14 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 43847 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| ELISA, Immunoprecipitation, Neutralization, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q9P0G3 | |
| Klk14 | |
| Mouse myeloma cell line NS0-derived recombinant human Kallikrein 14 Gln19-Met248, predicted. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction