Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kallikrein 15 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA14211

Catalog No. PA14211


Only null left
Add to Cart

Description

Description

PA1-4211 detects the Kallikrein-15 protein in human samples. PA1-4211 has been successfully used in Western blot procedures. The PA1-4211 immunogen is a synthetic peptide corresponding to residues H(75) N L R K R D G P E Q L R(87) of human Kallikrein-15.

Kallikreins are a subgroup of serine proteases and are implicated in carcinogenesis. A novel KLK gene, KLK12, is expressed in a variety of tissues including salivary gland, stomach, uterus, lung, thymus, prostate, colon, brain, thyroid, and trachea. It has applications as a novel cancer biomarker.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Kallikrein 15
Polyclonal
Unconjugated
KLK15
ACO; ACO protease; HSRNASPH; kallikrein 15; kallikrein related peptidase 15; kallikrein related-peptidase 15; kallikrein-15; kallikrein-like serine protease; kallikrein-related peptidase 15; KLK15; KLK-15; prostinogen
Rabbit
Antigen affinity chromatography
RUO
55554
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q9H2R5
KLK15
Synthetic peptide corresponding to residues H(75) N L R K R D G P E Q L R(87) of human Kallikrein-15.
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.