Learn More
Invitrogen™ Kallikrein 15 Polyclonal Antibody

Description
In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) Kallikrein 2, rhKallikrein 8, and rhKLK11 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | Kallikrein 15 |
| Applications | Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | KLK15 |
| Gene Accession No. | Q9H2R5 |
| Gene Alias | ACO; ACO protease; HSRNASPH; kallikrein 15; kallikrein related peptidase 15; kallikrein related-peptidase 15; kallikrein-15; kallikrein-like serine protease; kallikrein-related peptidase 15; KLK15; KLK-15; prostinogen |
| Gene Symbols | KLK15 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.