Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Kallikrein 15 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596205
Description
Immunogen sequence: RDGPEQLRTT SRVIPHPRYE ARSHRNDIML LRLVQPARLN PQVRPAVLPT RCPHPGEACV VSGWGLVSHN EPGTAGSPRS QVSLPDTLHC ANISIISDTS C; Positive Samples: Mouse kidney, Rat kidney.
Kallikreins are a subgroup of serine proteases and are implicated in carcinogenesis. A novel KLK gene, KLK12, is expressed in a variety of tissues including salivary gland, stomach, uterus, lung, thymus, prostate, colon, brain, thyroid, and trachea. It has applications as a novel cancer biomarker.
Specifications
Kallikrein 15 | |
Polyclonal | |
Unconjugated | |
KLK15 | |
KLK15 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 80-180 of human KLK15 (NP_0599792). | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
ELISA, Western Blot | |
0.74 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
ACO; ACO protease; HSRNASPH; kallikrein 15; kallikrein related peptidase 15; kallikrein related-peptidase 15; kallikrein-15; kallikrein-like serine protease; kallikrein-related peptidase 15; KLK15; KLK-15; prostinogen | |
Rabbit | |
Affinity chromatography | |
RUO | |
292873, 317652 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction