Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kallikrein 2 Antibody (JNJ-69086420) [Alexa Fluor™ 647], Novus Biologicals™
SDP

Catalog No. NB780586 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB780586 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB780586 Supplier Novus Biologicals Supplier No. NBP328696AF647
Only null left
Add to Cart
Add to Cart

Human Monoclonal Antibody

Kallikrein 2 Monoclonal antibody specifically detects Kallikrein 2 in Human samples. It is validated for ELISA,Functional Assay

Specifications

Antigen Kallikrein 2
Applications ELISA, Functional Assay
Classification Monoclonal
Clone JNJ-69086420
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias EC 3.4.21, EC 3.4.21.35, glandular kallikrein 2, Glandular kallikrein-1, hGK-1, hK2, kallikrein 2, prostatic, kallikrein-2, kallikrein-related peptidase 2, KLK2, KLK2A2, MGC12201, Tissue kallikrein-2
Host Species Human
Immunogen KLK2/ Kallikrein 2
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3817
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.