Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kallikrein 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579566
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579566 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579566 Supplier Invitrogen™ Supplier No. PA579566
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human MCF-7 cell, human COLO-320 cell, human SK-OV-3 cell, human HepG2 cell. IHC: human prostatic cancer tissue.

Glandular kallikreins cleave Met-Lys and Arg-Ser bonds in kininogen to release Lys-bradykinin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kallikrein 2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene KLK2
Gene Accession No. P20151
Gene Alias glandular kallikrein 2; glandular kallikrein-1; hGK-1; hK2; kallikrein 2, prostatic; kallikrein related peptidase 2; kallikrein-2; kallikrein-related peptidase 2; KLK2; KLK2A2; MGC12201; Tissue kallikrein 2; Tissue kallikrein-2
Gene Symbols KLK2
Host Species Rabbit
Immunogen E. coli-derived human Kallikrein 2 recombinant protein (Position: I25-P261).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3817
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.