Learn More
Invitrogen™ Kallikrein 2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109453
Description
Immunogen sequence: IVGGWECEKH SQPWQVAVYS HGWAHCGGVL VHPQWVLTAA HCLKKNSQVW LGRHNLFEPE DTGQRVPVSH SFPHPLYNMS LLKHQSLRPD EDSSHDLMLL RLSEPAKITD VVKVLGLPTQ EPALGTTCYA SGWGSIEPEE FLRPRSLQCV SLHLLSNDMC ARAYSEKVTE FMLCAGLWTG GKDTCGGDSG GPLVCNGVLQ GITSWGPEPC ALPEKPAVYT KVVHYR.
Glandular kallikreins cleave Met-Lys and Arg-Ser bonds in kininogen to release Lys-bradykinin.
Specifications
| Kallikrein 2 | |
| Polyclonal | |
| Unconjugated | |
| KLK2 | |
| glandular kallikrein 2; glandular kallikrein-1; hGK-1; hK2; kallikrein 2, prostatic; kallikrein related peptidase 2; kallikrein-2; kallikrein-related peptidase 2; KLK2; KLK2A2; MGC12201; Tissue kallikrein 2; Tissue kallikrein-2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 3817 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.50 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P20151 | |
| KLK2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 25-250 of human KLK2 (NP_005542.1). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.