Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kallikrein 2 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA527208

Catalog No. PIPA527208


Only null left
Add to Cart

Description

Description

Recommended positive controls: Molt-4. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Glandular kallikreins cleave Met-Lys and Arg-Ser bonds in kininogen to release Lys-bradykinin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Kallikrein 2
Polyclonal
Unconjugated
KLK2
glandular kallikrein 2; glandular kallikrein-1; hGK-1; hK2; kallikrein 2, prostatic; kallikrein related peptidase 2; kallikrein-2; kallikrein-related peptidase 2; KLK2; KLK2A2; MGC12201; Tissue kallikrein 2; Tissue kallikrein-2
Rabbit
Antigen affinity chromatography
RUO
3817
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P20151
KLK2
Recombinant fragment corresponding to a region within amino acids 112 and 261 of Human Kallikrein 2.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.