Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kallikrein 2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 108121AP150UL
Description
Glandular kallikreins cleave Met-Lys and Arg-Ser bonds in kininogen to release Lys-bradykinin.Specifications
| Kallikrein 2 | |
| Polyclonal | |
| Unconjugated | |
| KLK2 | |
| Glandular kallikrein 1, hGK 1, hK2, Kallikrein 2, kallikrein related peptidase 2, KLK2, KLK2A2, Tissue kallikrein 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16603, 3817 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P20151 | |
| KLK2, Klk2-ps | |
| Kallikrein 2 Fusion Protein Ag1077 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction