Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kallikrein 3/PSA Antibody (103), DyLight 550

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289633R
Description
Kallikrein 3/PSA Monoclonal specifically detects Kallikrein 3/PSA in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Kallikrein 3/PSA | |
Monoclonal | |
DyLight 550 | |
APS seminin, EC 3.4.21, EC 3.4.21.77, Gamma-seminoprotein, hK3, kallikrein 3, (prostate specific antigen), Kallikrein-3, kallikrein-related peptidase 3, KLK2A1, P-30 antigen, prostate specific antigen, prostate-specific antigen, PSA, PSA semenogelase, Semenogelase, Seminin | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Kallikrein 3/PSA (P07288; Met1-Pro261). | |
0.1 mL | |
Cancer, Cellular Markers, Prostate Cancer, Tumor Biomarkers | |
354 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
103 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction