Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kallikrein 3/PSA Antibody (5A11E9), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP151541JF646
Description
Kallikrein 3/PSA Monoclonal antibody specifically detects Kallikrein 3/PSA in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Kallikrein 3/PSA | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
5A11E9 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
APS seminin, EC 3.4.21, EC 3.4.21.77, Gamma-seminoprotein, hK3, kallikrein 3, (prostate specific antigen), Kallikrein-3, kallikrein-related peptidase 3, KLK2A1, P-30 antigen, prostate specific antigen, prostate-specific antigen, PSA, PSA semenogelase, Semenogelase, Seminin | |
Purified recombinant fragment of Prostate Specific Antigen (amino acids 26-251) expressed in E. coli. [UniProt# P07288] | |
0.1 mL | |
Cancer, Cellular Markers, Prostate Cancer, Tumor Biomarkers | |
354 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction