Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kallikrein 4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572768
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572768 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572768 Supplier Invitrogen™ Supplier No. PA572768
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Kallikrein 4 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Kallikreins are a subgroup of serine proteases and are implicated in carcinogenesis. A novel Kallikrein gene Prostase/KLK-L1 (also known as KLK4) is expressed and is up-regulated by androgens and progestins. KLK-L1 may be involved in the pathogenesis and/or progression of prostate, breast, and possibly other malignancies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kallikrein 4
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene KLK4
Gene Accession No. Q9Y5K2
Gene Alias AI2A1; androgen-regulated message 1; ARM1; EMSP; Emsp1; enamel matrix serine protease 1; Enamel matrix serine proteinase 1; ESMP1; kallikrein; kallikrein 4 (prostase, enamel matrix, prostate); kallikrein related peptidase 4; kallikrein related-peptidase 4 (prostase, enamel matrix, prostate); kallikrein-4; Kallikrein-like protein 1; kallikrein-related peptidase 4; Klk4; KLK-4; KLKL1; KLK-L1; Prostase; protease, serine 17 (enamel matrix, prostate); Prss17; PSTS; serine protease 17
Gene Symbols KLK4
Host Species Rabbit
Immunogen KLK4 antibody for a 15 amino acid peptide near the center of human KLK4. The immunogen is within amino acids 80 - 130 of KLK4.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9622
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.