Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kallikrein 4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576973
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576973 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576973 Supplier Invitrogen™ Supplier No. PA576973
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

Kallikreins are a subgroup of serine proteases and are implicated in carcinogenesis. A novel Kallikrein gene Prostase/KLK-L1 (also known as KLK4) is expressed and is up-regulated by androgens and progestins. KLK-L1 may be involved in the pathogenesis and/or progression of prostate, breast, and possibly other malignancies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kallikrein 4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene KLK4
Gene Accession No. Q9Y5K2, Q9Z0M1
Gene Alias AI2A1; androgen-regulated message 1; ARM1; EMSP; Emsp1; enamel matrix serine protease 1; Enamel matrix serine proteinase 1; ESMP1; kallikrein; kallikrein 4 (prostase, enamel matrix, prostate); kallikrein related peptidase 4; kallikrein related-peptidase 4 (prostase, enamel matrix, prostate); kallikrein-4; Kallikrein-like protein 1; kallikrein-related peptidase 4; Klk4; KLK-4; KLKL1; KLK-L1; Prostase; protease, serine 17 (enamel matrix, prostate); Prss17; PSTS; serine protease 17
Gene Symbols KLK4
Host Species Rabbit
Immunogen Recombinant full length Human Kallikrein 4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 408210, 56640, 9622
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.