Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kallikrein 4/Prostase/EMSP1 Antibody (104), Alexa Fluor™ 350, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290008AF350
Description
Kallikrein 4/Prostase/EMSP1 Monoclonal specifically detects Kallikrein 4/Prostase/EMSP1 in Human samples. It is validated for ELISA.Specifications
Kallikrein 4/Prostase/EMSP1 | |
Monoclonal | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Human KLK1 Human KLK3 Human KLK7 Human KLK8 Human KLK11 Human KLK13 Human Factor X Human HGFA Human UPA-1 Human KLK6 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
104 | |
ELISA | |
AI2A1, ARM1, EC 3.4.21, EC 3.4.21.-, EMSP, EMSP1MGC116827, Enamel matrix serine proteinase 1, kallikrein 4 (prostase, enamel matrix, prostate), kallikrein-4, Kallikrein-like protein 1, kallikrein-related peptidase 4, KLK-L1MGC116828, Prostase, PRSS17enamel matrix serine protease 1, PSTSandrogen-regulated message 1, Serine protease 17 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Kallikrein 4/Prostase/EMSP1 (NP_004908.3; Met1-Ser254). | |
0.1 mL | |
Apoptosis, Cancer, Tumor Suppressors | |
9622 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction