Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kallikrein 4/Prostase/EMSP1 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299892DL594
Description
Kallikrein 4/Prostase/EMSP1 Polyclonal specifically detects Kallikrein 4/Prostase/EMSP1 in Human samples. It is validated for ELISA.Specifications
Kallikrein 4/Prostase/EMSP1 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
DyLight 594 | |
AI2A1, ARM1, EC 3.4.21, EC 3.4.21.-, EMSP, EMSP1MGC116827, Enamel matrix serine proteinase 1, kallikrein 4 (prostase, enamel matrix, prostate), kallikrein-4, Kallikrein-like protein 1, kallikrein-related peptidase 4, KLK-L1MGC116828, Prostase, PRSS17enamel matrix serine protease 1, PSTSandrogen-regulated message 1, Serine protease 17 | |
Produced in rabbits immunized with purified, recombinant Kallikrein 4/Prostase/EMSP1 (NP_004908.3; Met1-Ser254). | |
0.1 mL | |
Apoptosis, Cancer, Tumor Suppressors | |
9622 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction