Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kallikrein 5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582500
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582500 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582500 Supplier Invitrogen™ Supplier No. PA582500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CDHPSNTVPS GSNQDLGAGA GEDARSDDSS SRIINGSDCD MHTQPWQAAL LLRPNQLYCG AVLVHPQWLL TAAHCRKKVF RVRLGHYSLS PVYESGQQMF QGVKSIPHPG YSHPGHSNDL MLIKLNRRIR PTKDVRPINV SSH Highest antigen sequence indentity to the following orthologs: Rat - 72%, Mouse - 72%.

Kallikreins are a subgroup of serine proteases and are implicated in carcinogenesis. A novel KLK gene, KLK12, is expressed in a variety of tissues including salivary gland, stomach, uterus, lung, thymus, prostate, colon, brain, thyroid, and trachea. It has applications as a novel cancer biomarker.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kallikrein 5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Klk5
Gene Accession No. Q9Y337
Gene Alias 1110030O19Rik; HK5; kallikrein 5; Kallikrein L2; Kallikrein like 2; kallikrein related peptidase 5; kallikrein related-peptidase 5; Kallikrein5; kallikrein-5; kallikrein-5-like; kallikrein-like protein 2; kallikrein-related peptidase 5; KLK 5; KLK L2; KLK5; KLK-5; KLKL 2; KLKL2; KLK-L2; LOC102546758; SCTE; Stratum corneum tryptic enzyme; UNQ570/PRO1132
Gene Symbols Klk5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KLK5. Recombinant protein control fragment (Product #RP-90432).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25818
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.