Learn More
Invitrogen™ Kallikrein 5 Polyclonal Antibody

Description
PA1-21722 detects Kallikrein-L2 in Human samples.
Specifications
Specifications
| Antigen | Kallikrein 5 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | Klk5 |
| Gene Accession No. | Q9Y337 |
| Gene Alias | 1110030O19Rik; HK5; kallikrein 5; Kallikrein L2; Kallikrein like 2; kallikrein related peptidase 5; kallikrein related-peptidase 5; Kallikrein5; kallikrein-5; kallikrein-5-like; kallikrein-like protein 2; kallikrein-related peptidase 5; KLK 5; KLK L2; KLK5; KLK-5; KLKL 2; KLKL2; KLK-L2; LOC102546758; SCTE; Stratum corneum tryptic enzyme; UNQ570/PRO1132 |
| Gene Symbols | Klk5 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.