Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kallikrein 6/Neurosin Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298247AF750
Description
Kallikrein 6/Neurosin Polyclonal specifically detects Kallikrein 6/Neurosin in Mouse samples. It is validated for Western Blot, ELISA.Specifications
Kallikrein 6/Neurosin | |
Polyclonal | |
Western Blot, ELISA | |
Bssp, EC 3.4.21, EC 3.4.21.-, hK6, kallikrein 6 (neurosin, zyme), kallikrein-6, kallikrein-related peptidase 6, Klk7, neurosin, Protease M, protease, serine, 18, PRSS18, PRSS9MGC9355, Serine protease 18, Serine protease 9, SP59, Zyme | |
Produced in rabbits immunized with purified, recombinant Kallikrein 6/Neurosin (NP_035307.2; Met 8-Leu 253). | |
0.1 mL | |
Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Neurodegeneration, Neuroscience, Signal Transduction | |
5653 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction