Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kallikrein 6/Neurosin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | Kallikrein 6/Neurosin |
---|---|
Dilution | Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:20 |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP106497SS
![]() |
Novus Biologicals
NBP106497SS |
0.025 mL |
Each for $181.90
|
|
|||||
NBP106497
![]() |
Novus Biologicals
NBP106497 |
0.1 mL |
Each for $487.50
|
|
|||||
Description
Kallikrein 6/Neurosin Polyclonal specifically detects Kallikrein 6/Neurosin in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Kallikrein 6/Neurosin | |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Endothelial Cell Markers, Extracellular Matrix, Neurodegeneration, Neuroscience, Signal Transduction | |
Whole antisera with 0.1% Sodium Azide | |
Bssp, EC 3.4.21, EC 3.4.21.-, hK6, kallikrein 6 (neurosin, zyme), kallikrein-6, kallikrein-related peptidase 6, Klk7, neurosin, Protease M, protease, serine, 18, PRSS18, PRSS9MGC9355, Serine protease 18, Serine protease 9, SP59, Zyme | |
KLK6 | |
IgG | |
Unpurified | |
Human. Immunogen sequence has 85% identity with canine, 82% identity with horse and 80% identity with bovine. |
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:20 | |
Polyclonal | |
Antisera | |
RUO | |
Human | |
Q92876 | |
5653 | |
Full length human Kallikrein 6 protein expressed in E. coli. [Swiss-Prot# Q92876] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
38 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title