Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kallikrein 6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588324
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588324 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588324 Supplier Invitrogen™ Supplier No. PA588324
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AASHDQDIML LRLARPAKLS ELIQPLPLER DCSANTTSCH ILGWGKTADG DFPDTIQCAY IHLVSREECE HAYPGQITQN MLCAGDEKYG KDSCQGDSGG PLVCGDHLRG LVSWGNIPCG SKEKPGVYTN VCRYTNWIQK TIQAK; Positive Samples: HT-29; Cellular Location: Cytoplasm, Microsome, Mitochondrion, Nucleus, nucleolus.

Kallikreins, including Kallikrein 6, are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. The KLK6 enzyme is regulated by steroid hormones. In tissue culture, the enzyme has been found to generate amyloidogenic fragments from the amyloid precursor protein, suggesting a potential for involvement in Alzheimer's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kallikrein 6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Klk6
Gene Accession No. Q92876
Gene Alias 0610007D04Rik; AI849898; brain serine protease; BSP; Bssp; Glandular kallikrein K1; hK6; Kal; KAL-B; kallikrein 1; kallikrein 29; kallikrein 6; kallikrein 6 (neurosin); kallikrein 6 (neurosin, zyme); kallikrein 7; kallikrein related peptidase 6; kallikrein related-peptidase 6; kallikrein renal/pancreas/salivary; kallikrein-1; Kallikrein6; Kallikrein-6; Klk1; Klk1b6; Klk29; Klk6; Klk-6; Klk7; MGC9355; mGK-6; MSP; myelencephalon-specific protease; myofibril-bound protease; neurosin; Protease M; protease serine 9 (neurosin); protease, serine, 18; protease, serine, 9; PRSS18; PRSS9; renal kallikrein; Serine protease 18; serine protease 9; SP59; tissue kallikrein-6; true tissue kallikrein; zyme
Gene Symbols Klk6
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 100 to the C-terminus of human KLK6 (NP_0027651).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5653
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.