Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kallikrein 7 Antibody (MM0433-5J28), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP211753AF750
Description
Kallikrein 7 Monoclonal specifically detects Kallikrein 7 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Kallikrein 7 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
KLK7 | |
Human recombinant Kallikrein 7 | |
Primary | |
Human | |
IgG2 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
MM0433-5J28 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
EC 3.4.21, EC 3.4.21.117, hK7, hSCCE, kallikrein 7 (chymotryptic, stratum corneum), kallikrein-related peptidase 7, protease, serine, 6, PRSS6, SCCEkallikrein-7, Serine protease 6, signal protein, Stratum corneum chymotryptic enzyme | |
Mouse | |
0.1 mL | |
5650 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction