Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kallikrein 7 (KLK7) Mouse anti-Human, Unconjugated, Clone: 83-1, MilliporeSigma™

Catalog No. MABN243025 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN2430-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN2430-25 Supplier MilliporeSigma Supplier No. MABN243025UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Kallikrein 7 (KLK7), clone 83-1, Cat. No. MABN2430, is a highly specific mouse monoclonal antibody that targets Kallikrein-7 and has been tested in ELISA and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kallikrein 7 (KLK7)
Applications Western Blot, ELISA
Classification Monoclonal
Clone 83-1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Sodium bicarbonate buffer, pH 8.2 with 0.02% sodium azide.
Gene Alias Serine protease 6;EC 3.4.21.117;hK7;Stratum corneum chymotryptic enzyme;hSCCE
Gene Symbols KLK7;PRSS6;SCCE
Host Species Mouse
Immunogen Full-length human recombinant Kallikrein-7.
Molecular Weight of Antigen ∽27 kDa observed; 27.53 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Quantity 25 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity Clone 83-1 detects Kallikrein -7 in human tissues.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.