Learn More
Invitrogen™ Kallikrein 8 Monoclonal Antibody (369910)

Description
In direct ELISAs, no cross-reactivity with recombinant human (rh) UPA-1, rhKallikrein 1, 2, 3, 4, 5, 7, 9, 10, 11, 12, 13, 14, 15, rhKallikrein B1, rhHGFA, rhFactor VII, rhFactor X, rhFactor XI, rhProc, or rhThrombin is observed. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
Specifications
Specifications
| Antigen | Kallikrein 8 |
| Applications | Immunoprecipitation, Neutralization |
| Classification | Monoclonal |
| Clone | 369910 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | KLK8 |
| Gene Accession No. | O60259 |
| Gene Alias | brain serine protease 1; BSP1; EGK_10950; H NRPN; hK8; HNP; kallikrein 8; kallikrein 8 (neuropsin/ovasin); kallikrein related peptidase 8; kallikrein related-peptidase 8; Kallikrein8; kallikrein-8; kallikrein-related peptidase 8; Klk8; mK8; neuropsin; NP; Nrpn; Ovasin; PRO322; protease, serine, 19 (neuropsin); Prss19; Serine protease 19; serine protease TADG-14; TADG14; Tumor-associated differentially expressed gene 14 protein; UNQ283; UNQ283/PRO322 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.