Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Kallikrein 8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557388
Description
Immunogen sequence: GSKVKPISLA DHCTQPGQKC TVSGWGTVTS PRENFPDTLN CAEVKIFPQK KCEDAYPGQI TDGMVCAGSS KGADTCEGDS GG Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 77%.
Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in tandem in a gene cluster on chromosome 19. The encoded protein may be involved in proteolytic cascade in the skin and may serve as a biomarker for ovarian cancer. Alternate splicing of this gene results in multiple transcript variants encoding different isoforms.
Specifications
Kallikrein 8 | |
Polyclonal | |
Unconjugated | |
KLK8 | |
brain serine protease 1; BSP1; EGK_10950; H NRPN; hK8; HNP; kallikrein 8; kallikrein 8 (neuropsin/ovasin); kallikrein related peptidase 8; kallikrein related-peptidase 8; Kallikrein8; kallikrein-8; kallikrein-related peptidase 8; Klk8; mK8; neuropsin; NP; Nrpn; Ovasin; PRO322; protease, serine, 19 (neuropsin); Prss19; Serine protease 19; serine protease TADG-14; TADG14; Tumor-associated differentially expressed gene 14 protein; UNQ283; UNQ283/PRO322 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11202 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60259 | |
KLK8 | |
Recombinant protein corresponding to Human Kallikrein 8. Recombinant protein control fragment (Product #RP-102678). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction