Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kallikrein 9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547497
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547497 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547497 Supplier Invitrogen™ Supplier No. PA547497
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human KLK-1, -3, -4, -5, -6, -7, -8, -10, -11, -12, -13, -14, and -15 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. KLK9 is a novel kallikrein with potential application for diagnosis, monitoring and therapeutics of various cancers including those of the breast, prostate and testis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kallikrein 9
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene KLK9
Gene Accession No. Q9UKQ9
Gene Alias 1200016C12Rik; AI324041; kallikrein 9; kallikrein related peptidase 9; kallikrein related-peptidase 9; kallikrein-9; Kallikrein-like protein 3; kallikrein-related peptidase 9; KLK9; KLK-9; KLKL3; KLK-L3
Gene Symbols KLK9
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Kallikrein 9 Asp20-Asn250.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284366
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.