Learn More
Invitrogen™ Kallikrein 9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595062
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: MCF-7 Cell, A431 Cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. KLK9 is a novel kallikrein with potential application for diagnosis, monitoring and therapeutics of various cancers including those of the breast, prostate and testis.
Specifications
Kallikrein 9 | |
Polyclonal | |
Unconjugated | |
KLK9 | |
1200016C12Rik; AI324041; kallikrein 9; kallikrein related peptidase 9; kallikrein related-peptidase 9; kallikrein-9; Kallikrein-like protein 3; kallikrein-related peptidase 9; KLK9; KLK-9; KLKL3; KLK-L3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
284366 | |
-20°C | |
Lyophilized |
Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
Q9UKQ9 | |
KLK9 | |
A synthetic peptide corresponding to a sequence at the C-terminus of human Kallikrein 9 (236-250aa SVCHYLDWIQEIMEN). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.