Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Kallikrein 9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115443
Description
Antibody detects endogenous levels of total Kallikrein 9.
Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. KLK9 is a novel kallikrein with potential application for diagnosis, monitoring and therapeutics of various cancers including those of the breast, prostate and testis.
Specifications
| Kallikrein 9 | |
| Polyclonal | |
| Unconjugated | |
| KLK9 | |
| 1200016C12Rik; AI324041; kallikrein 9; kallikrein related peptidase 9; kallikrein related-peptidase 9; kallikrein-9; Kallikrein-like protein 3; kallikrein-related peptidase 9; KLK9; KLK-9; KLKL3; KLK-L3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 284366, 292851 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P07647, Q9UKQ9 | |
| KLK9 | |
| A synthesized peptide derived from human KLK9(Accession Q9UKQ9), corresponding to amino acid residues C200-N250. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction