Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kanadaptin Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306068C
Description
Kanadaptin Polyclonal specifically detects Kanadaptin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Kanadaptin | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
FLJ10624, FLJ41004, HLC3, HLC-3, Human lung cancer oncogene 3 protein, kanadaptin, Kidney anion exchanger adapter protein, kidney anion exchanger adaptor protein, lung cancer oncogene 3, MGC120646, MGC120648, solute carrier family 4 (anion exchanger), member 1, adapter protein, solute carrier family 4 (anion exchanger), member 1, adaptor protein, Solute carrier family 4 anion exchanger member 1 adapter protein | |
Produced in rabbits immunized with E. coli-derived Human Kanadaptin fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
22950 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction