Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kanamycin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA175225
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA175225 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA175225 Supplier Invitrogen™ Supplier No. PA175225
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Working Concentration: 0.3125 μg/mL. Sensitivity: 10 ng/mL Kanamycin produces 57% inhibition in a competitive ELISA employing Kanamycin polyclonal antibody.

Kanamycin is only found in individuals that have used or taken this drug. It is an antibiotic complex produced by Streptomyces kanamyceticus from Japanese soil. Comprises 3 components: kanamycin A, the major component, and kanamycins B and C, the minor components. [PubChem]Aminoglycosides like kanamycin irreversibly bind to specific 30S-subunit proteins and 16S rRNA. Specifically Kanamycin binds to four nucleotides of 16S rRNA and a single amino acid of protein S12. This interferes with decoding site in the vicinity of nucleotide 1400 in 16S rRNA of 30S subunit. This region interacts with the wobble base in the anticodon of tRNA. This leads to interference with the initiation complex, misreading of mRNA so incorrect amino acids are inserted into the polypeptide leading to nonfunctional or toxic peptides and the breakup of polysomes into nonfunctional monosomes. [National Center for Biotechnology Information. PubChem Compound Database; CID=6032].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kanamycin
Applications ELISA
Classification Polyclonal
Concentration 14.042 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Alias C18H36N4O11; Kanamicina; Kanamycin A; Kanamycin Base; Kanamycinum
Host Species Sheep
Immunogen Kanamycin-BTG.
Purification Method Purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.