Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KANK1 Monoclonal Antibody (1773CT634.49.61)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA537733
Description
KANK1 Monoclonal Antibody for Western Blot, IHC (P)
Ankyrins are membrane adaptor molecules that play important roles in the control of cytoskeleton formation by regulating actin polymerization. KANK1 (KN motif and ankyrin repeat domain-containing protein 1), also known as ANKRD15, is a 1,352 amino acid protein that contains at least 12 exons and 5 ANK repeats. It binds to beta-catenin and regulates its subcellular distribution. KANK1 is ubiquitously expressed and localizes to cytoplasm. It may function as a tumor suppressor for renal cell carcinoma. Mutations in this gene cause cerebral palsy spastic quadriplegic type 2, a central nervous system development disorder.
Specifications
KANK1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q14678 | |
Kank1 | |
recombinant protein of human KANK1. | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Immunohistochemistry (Paraffin), Western Blot | |
1773CT634.49.61 | |
Unconjugated | |
Kank1 | |
A930031B09Rik; ANKRD15; ankyrin repeat domain 15; ankyrin repeat domain-containing protein 15; AU015049; AW121052; CPSQ2; D330024H06Rik; KANK; Kank1; KIAA0172; Kidney ankyrin repeat-containing protein; KN motif and ankyrin repeat domain-containing protein 1; KN motif and ankyrin repeat domains 1; mKIAA0172 | |
Mouse | |
Protein G | |
RUO | |
23189 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction