Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KANK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552159
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552159 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552159 Supplier Invitrogen™ Supplier No. PA552159
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: INVCGVRKRS YSAGNASQLE QLSRARRSGG ELYIDYEEEE METVEQSTQR IKEFRQLTAD MQALEQKIQD SSCEASSELR ENGECRSVAV GAEENMNDIV VYHRGSRSCK DAAVGTLVEM RNCGVSVTEA MLGVMTEADK EIE Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 81%.

Ankyrins are membrane adaptor molecules that play important roles in the control of cytoskeleton formation by regulating actin polymerization. KANK1 (KN motif and ankyrin repeat domain-containing protein 1), also known as ANKRD15, is a 1,352 amino acid protein that contains at least 12 exons and 5 ANK repeats. It binds to beta-catenin and regulates its subcellular distribution. KANK1 is ubiquitously expressed and localizes to cytoplasm. It may function as a tumor suppressor for renal cell carcinoma. Mutations in this gene cause cerebral palsy spastic quadriplegic type 2, a central nervous system development disorder.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KANK1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Kank1
Gene Accession No. Q14678
Gene Alias A930031B09Rik; ANKRD15; ankyrin repeat domain 15; ankyrin repeat domain-containing protein 15; AU015049; AW121052; CPSQ2; D330024H06Rik; KANK; Kank1; KIAA0172; Kidney ankyrin repeat-containing protein; KN motif and ankyrin repeat domain-containing protein 1; KN motif and ankyrin repeat domains 1; mKIAA0172
Gene Symbols Kank1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KANK1. Recombinant protein control fragment (Product #RP-101071).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23189
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.