Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KANK1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565999
Description
Immunogen sequence: APLGMMTGLDH YIERIQKLLA EQQTLLAENY SELAEAFGEP HSQMGSLNSQ LISTLSSINS VMKSASTEEL RNPDFQKTSL GKITGNYLGY TCKCGGLQSG SPLSSQTSQP EQEVGTSEGK PISSLDAFPT QEGTLSPVNL TDDQ Highest antigen sequence identity to the following orthologs - mouse 77%, rat 79%.
Ankyrins are membrane adaptor molecules that play important roles in the control of cytoskeleton formation by regulating actin polymerization. KANK1 (KN motif and ankyrin repeat domain-containing protein 1), also known as ANKRD15, is a 1,352 amino acid protein that contains at least 12 exons and 5 ANK repeats. It binds to beta-catenin and regulates its subcellular distribution. KANK1 is ubiquitously expressed and localizes to cytoplasm. It may function as a tumor suppressor for renal cell carcinoma. Mutations in this gene cause cerebral palsy spastic quadriplegic type 2, a central nervous system development disorder.
Specifications
KANK1 | |
Polyclonal | |
Unconjugated | |
Kank1 | |
A930031B09Rik; ANKRD15; ankyrin repeat domain 15; ankyrin repeat domain-containing protein 15; AU015049; AW121052; CPSQ2; D330024H06Rik; KANK; Kank1; KIAA0172; Kidney ankyrin repeat-containing protein; KN motif and ankyrin repeat domain-containing protein 1; KN motif and ankyrin repeat domains 1; mKIAA0172 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23189 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q14678 | |
Kank1 | |
Recombinant Human KANK1. Recombinant protein control fragment (Product #RP-106684). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction