Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KANK2 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10068217AF700
Description
KANK2 Polyclonal antibody specifically detects KANK2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
KANK2 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
ANKRD25, ankyrin repeat domain 25, Ankyrin repeat domain-containing protein 25, DKFZp434N161, FLJ20004, KIAA1518SRC1-interacting protein, kidney ankyrin repeat-containing protein 2, KN motif and ankyrin repeat domain-containing protein 2, KN motif and ankyrin repeat domains 2, Matrix-remodeling-associated protein 3, matrix-remodelling associated 3, MGC119707, MXRA3, SIP, SRC-1 interacting protein, SRC-1-interacting protein | |
The immunogen recognized by this antibody maps to a region between residue 550 and 600 of human KN motif and ankyrin repeat domains 2 (SRC-1 interacting protein) using the numbering given in entry NP_056308.2 (GeneID 25959). | |
0.1 mL | |
Cancer, Cell Cycle and Replication | |
25959 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction