Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KANK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116620
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116620 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116620 Supplier Invitrogen™ Supplier No. PA5116620
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse skeletal muscle, Mouse heart, Mouse kidney Immunogen sequence: MAQVLHVPAP FPGTPGPASP PAFPAKDPDP PYSVETPYGY RLDLDFLKYV DDIEKGHTLR RVAVQRRPRL SSLPRGPGSW WTSTESLCSN ASGDSRHSAY SYCGRGFYPQ YGALETRGGF NPRVERTLLD ARRRLEDQAA TPTGLGSLTP SAAGSTASLV GVGLPPPTPR SSGLSTPVPP SAGHLAHVRE QMAGALRKLR QLEEQVKLIP VLQVKLSVLQ EEKRQLTVQL KSQKFLGHPT AGRGRSELCL DLPDPPEDPV ALETRSVGTW VRERDLGMPD GEAALAAKVA VLETQLKKAL.

This gene encodes a member of the KN motif and ankyrin repeat domains (KANK) family of proteins, which play a role in cytoskeletal formation by regulating actin polymerization. The encoded protein functions in the sequestration of steroid receptor coactivators and possibly other proteins. Mutations in this gene are associated with impaired kidney podocyte function and nephrotic syndrome, and keratoderma and woolly hair.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KANK2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene KANK2
Gene Accession No. Q63ZY3, Q8BX02
Gene Alias AI504612; ANKRD25; ankyrin repeat domain 25; ankyrin repeat domain-containing protein 25; BC010245; Kank2; KIAA1518; kidney ankyrin repeat-containing protein 2; KN motif and ankyrin repeat domain-containing protein 2; KN motif and ankyrin repeat domains 2; matrix-remodeling-associated protein 3; MXRA3; PPKWH; RGD1563532; SIP; SRC-1-interacting protein; SRC1-interacting protein; SRC-interacting protein
Gene Symbols KANK2
Host Species Rabbit
Immunogen Recombinant Protein (or fragment).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100361376, 235041, 25959
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.