Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KANK4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553113
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553113 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553113 Supplier Invitrogen™ Supplier No. PA553113
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IKAREQRIRE LEFTVAQLEG QFHQENAKDT QGQTDVMVNT DPVHGLLTRE SCDKGIEVNL LGSMESESWG HRGEENGLLW GPDGHKQGNQ SPAERVLLPQ LSLPQGPEQV LTSSVHSFLS TELRIEEAGT EQEGGPQGGT RGAGGFLWGS Highest antigen sequence identity to the following orthologs: Mouse - 54%, Rat - 55%.

KANK4 is a protein coding gene that may be involved in the control of cytoskeleton formation by regulating actin polymerization. Diseases associated with KANK4 include Fuchs' Endothelial Dystrophy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KANK4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KANK4
Gene Accession No. Q5T7N3
Gene Alias ANKRD38; ankyrin repeat domain 38; ankyrin repeat domain-containing protein 38; dJ1078M7.1; KANK4; kidney ankyrin repeat-containing protein 4; KN motif and ankyrin repeat domain-containing protein 4; KN motif and ankyrin repeat domains 4
Gene Symbols KANK4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KANK4. Recombinant protein control fragment (Product #RP-90760).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 163782
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.