Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KANSL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552356
Description
Immunogen sequence: APLLERLSQL DSCVHPVLAF PDDVPTSLHF QSMLKSQWQN KPFDKIKPPK KLSLKHRAPM PGSLPDSARK DRHKLVSSFL TTAKLSHHQT RPDRTHRQHL DDVGAVPMVE RVTAPKAERL LNPPPPVHDP NHSKMRLRDH SS Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 55%.
This gene encodes a nuclear protein that is a subunit of two protein complexes involved with histone acetylation, the MLL1 complex and the NSL1 complex. The corresponding protein in Drosophila interacts with K(lysine) acetyltransferase 8, which is also a subunit of both the MLL1 and NSL1 complexes.
Specifications
KANSL1 | |
Polyclonal | |
Unconjugated | |
KANSL1 | |
1700081L11Rik; 9430041J06Rik; CENP-36; centromere protein 36; centromere protein 36 antibody; DKFZp686P06109 antibody; DKFZp727C091 antibody; hMSL1v1; Kansl1; KAT8 regulatory NSL complex subunit 1; KAT8 regulatory NSL complex subunit 1 antibody; KDVS; KIAA1267; KIAA1267 antibody; male-specific lethal 1 homolog; MGC102843 antibody; mKIAA1267; MLL1/MLL complex subunit KANSL1; MLL1/MLL complex subunit male-specific lethal 1 homolog antibody; MSL1 homolog 1; MSL1V1; MSL1v1 antibody; non-specific lethal 1 homolog; NSL complex protein NSL1; NSL1; RGD1311429 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
284058 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z3B3 | |
KANSL1 | |
Recombinant protein corresponding to Human KANSL1. Recombinant protein control fragment (Product #RP-89716). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction